Learn More
Meso Scale Discovery R-PLEX Human proMMP-13 ASY
SDP
Catalog No. 5000079489
Encompass
The R-Plex Human ProMMP-13 Assay is an immunoassay kit for measuring ProMMP-13 in biological fluids It uses a quantitative sandwich enzyme immunoassay and colorimetric detection at 450nm Sensitivity is 0 3 ng/mL with a range of 0 78-50 ng/mL The kit includes a 96-well plate standard diluent biotinylated antibody avidin-HRP conjugate substrate and stop solution Total assay time is about 3 hours Suitable for ProMMP-13 research it requires an ELISA plate reader
Catalog No. 50-000-79489 Supplier Meso Scale Discovery Supplier No. K151ABER2
The R-Plex Human ProMMP-13 Assay is an immunoassay kit for measuring ProMMP-13 in biological fluids It uses a quantitative sandwich enzyme immunoassay and colorimetric detection at 450nm Sensitivity is 0 3 ng/mL with a range of 0 78-50 ng/mL The kit includes a 96-well plate standard diluent biotinylated antibody avidin-HRP conjugate substrate and stop solution Total assay time is about 3 hours Suitable for ProMMP-13 research it requires an ELISA plate reader