Learn More
Meso Scale Discovery R-PLEX Human proMMP-13 ASYQP
SDP
Catalog No. 5000079483
Encompass
The R-Plex Human ProMMP-13 Assay is an immunoassay kit for the quantitative measurement of proMMP-13 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) technology on the Meso Scale Discovery (MSD) platform The kit includes a 96-well plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG calibrators and controls The assay has a range of detection from 0 05-1000 ng/mL and takes approximately 3 5 hours to complete The kit provides sensitive accurate and reproducible quantitation of proMMP-13
Catalog No. 50-000-79483 Supplier Meso Scale Discovery Supplier No. K151ABER22
Add to Cart
Add to Cart
The R-Plex Human ProMMP-13 Assay is an immunoassay kit for the quantitative measurement of proMMP-13 in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) technology on the Meso Scale Discovery (MSD) platform The kit includes a 96-well plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG calibrators and controls The assay has a range of detection from 0 05-1000 ng/mL and takes approximately 3 5 hours to complete The kit provides sensitive accurate and reproducible quantitation of proMMP-13