Learn More
Meso Scale Discovery R-PLEX Human PSP-D ASY
SDP
Catalog No. 5000079070
Encompass
The R-PLEX Human PSP-D Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring Prostate Specific Protein (PSP) and Prostate Specific Antigen (PSA) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results This research-use-only assay aids in studying PSP and PSA levels for prostate cancer progression and treatment monitoring The kit includes a 96-well plate antibodies calibrators controls diluent read buffer and wash buffer for one plate
Catalog No. 50-000-79070 Supplier Meso Scale Discovery Supplier No. K1519XR2
The R-PLEX Human PSP-D Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring Prostate Specific Protein (PSP) and Prostate Specific Antigen (PSA) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results This research-use-only assay aids in studying PSP and PSA levels for prostate cancer progression and treatment monitoring The kit includes a 96-well plate antibodies calibrators controls diluent read buffer and wash buffer for one plate