Learn More
Meso Scale Discovery R-PLEX Human PTEN total ASY
SDP
Catalog No. 5000081623
Encompass
The R-PLEX Human PTEN Total Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of total PTEN protein in human cell and tissue lysates The assay uses an electrochemiluminescent detection method and provides high sensitivity a broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls to generate a standard curve and validate assay performance The assay takes approximately 4 hours to complete and is suitable for research use only
Catalog No. 50-000-81623 Supplier Meso Scale Discovery Supplier No. K151AKVR2
The R-PLEX Human PTEN Total Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of total PTEN protein in human cell and tissue lysates The assay uses an electrochemiluminescent detection method and provides high sensitivity a broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls to generate a standard curve and validate assay performance The assay takes approximately 4 hours to complete and is suitable for research use only