Learn More
Meso Scale Discovery R-PLEX Human PYY active ASY
SDP
Catalog No. 5000078851
Encompass
The R-PLEX Hu PYY Active Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active PYY levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The active assay specifically measures the biologically active form of PYY and allows sensitive reproducible quantitation of PYY levels
Catalog No. 50-000-78851 Supplier Meso Scale Discovery Supplier No. K1513RR2
The R-PLEX Hu PYY Active Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active PYY levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The active assay specifically measures the biologically active form of PYY and allows sensitive reproducible quantitation of PYY levels