Learn More
Meso Scale Discovery R-PLEX Human Relaxin2 ASY
SDP
Catalog No. 5000079455
Encompass
The R-PLEX Human Relaxin-2 Assay Kit from Meso Scale Discovery is an immunoassay that quantitatively measures human relaxin-2 in serum plasma cell culture supernatants and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of relaxin-2 with a lower limit of detection of 0 08 ng/mL and an assay time of just 3 5 hours The kit includes a 96-well plate pre-coated with capture antibody detection antibody calibrator and assay diluent optimized for relaxin-2 detection This assay allows high-throughput quantification of relaxin-2 levels with accuracy and precision
Catalog No. 50-000-79455 Supplier Meso Scale Discovery Supplier No. K151ABFR2
The R-PLEX Human Relaxin-2 Assay Kit from Meso Scale Discovery is an immunoassay that quantitatively measures human relaxin-2 in serum plasma cell culture supernatants and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of relaxin-2 with a lower limit of detection of 0 08 ng/mL and an assay time of just 3 5 hours The kit includes a 96-well plate pre-coated with capture antibody detection antibody calibrator and assay diluent optimized for relaxin-2 detection This assay allows high-throughput quantification of relaxin-2 levels with accuracy and precision