Learn More
Meso Scale Discovery R-PLEX Human Relaxin2 ASYQP
SDP
Catalog No. 5000078854
Encompass
The R-PLEX Human Relaxin-2 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human relaxin-2 in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a sensitive and rapid method for detecting relaxin-2 with a lower limit of detection of 0 08 ng/mL and an assay time of just 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to complete the assay
Catalog No. 50-000-78854 Supplier Meso Scale Discovery Supplier No. K151ABFR22
The R-PLEX Human Relaxin-2 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human relaxin-2 in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a sensitive and rapid method for detecting relaxin-2 with a lower limit of detection of 0 08 ng/mL and an assay time of just 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to complete the assay