Learn More
Meso Scale Discovery R-PLEX Human Renin ASY
SDP
Catalog No. 5000079484
Encompass
The R-PLEX Human Renin Assay from Meso Scale Discovery is a multiplex immunoassay that measures active renin in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time The R-PLEX assay quantitatively measures renin levels and is designed for use in biomarker research drug discovery and development This product requires the MSD QuickPlex SQ120 instrument and provides components for 96 tests in the kit
Catalog No. 50-000-79484 Supplier Meso Scale Discovery Supplier No. K151ABZR2
The R-PLEX Human Renin Assay from Meso Scale Discovery is a multiplex immunoassay that measures active renin in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 2 5 hour turnaround time The R-PLEX assay quantitatively measures renin levels and is designed for use in biomarker research drug discovery and development This product requires the MSD QuickPlex SQ120 instrument and provides components for 96 tests in the kit