Learn More
Meso Scale Discovery R-PLEX Human Resistin ASYQP
SDP
Catalog No. 5000080485
Encompass
The Meso Scale Discovery R-PLEX Human Resistin Assay Kit is a multiplex immunoassay for measuring resistin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It aids research on resistin s role in obesity inflammation diabetes and other diseases
Catalog No. 50-000-80485 Supplier Meso Scale Discovery Supplier No. K151ZOR22
The Meso Scale Discovery R-PLEX Human Resistin Assay Kit is a multiplex immunoassay for measuring resistin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It aids research on resistin s role in obesity inflammation diabetes and other diseases