Learn More
Meso Scale Discovery R-PLEX Human S100A8/MRP8 ASY
SDP
Catalog No. 5000078892
Encompass
The R-PLEX Human S100A8 (MRP8) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures S100A8 protein levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It is for research use only
Catalog No. 50-000-78892 Supplier Meso Scale Discovery Supplier No. K1510PR2
The R-PLEX Human S100A8 (MRP8) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures S100A8 protein levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It is for research use only