Learn More
Meso Scale Discovery R-PLEX Human S100B ASY
SDP
Catalog No. 5000078392
Encompass
The Meso Scale Discovery R-PLEX Human S100B Assay QuickPlex 5-PL Kit is a multiplex immunoassay for measuring S100B protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive seals for one 96-well plate It quantifies S100B levels from 0 04 ng/mL to 1 000 ng/mL requires 50 L of sample and is compatible with Meso Scale Discovery SECTOR Imagers
Catalog No. 50-000-78392 Supplier Meso Scale Discovery Supplier No. K1512ER2
The Meso Scale Discovery R-PLEX Human S100B Assay QuickPlex 5-PL Kit is a multiplex immunoassay for measuring S100B protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive seals for one 96-well plate It quantifies S100B levels from 0 04 ng/mL to 1 000 ng/mL requires 50 L of sample and is compatible with Meso Scale Discovery SECTOR Imagers