Learn More
Meso Scale Discovery R-PLEX Human S100B ASYQP
SDP
Catalog No. 5000080285
Encompass
The Meso Scale Discovery R-PLEX Human S100B Assay QuickPlex Kit is a multiplex immunoassay that quantitatively measures S100B protein in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 5-plate kit includes all necessary reagents and plates for analyzing S100B levels across multiple samples With minimal sample volume requirements and a streamlined workflow this assay kit enables efficient profiling of S100B which plays a role in diseases involving the central nervous system
Catalog No. 50-000-80285 Supplier Meso Scale Discovery Supplier No. K1512ER22
The Meso Scale Discovery R-PLEX Human S100B Assay QuickPlex Kit is a multiplex immunoassay that quantitatively measures S100B protein in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 5-plate kit includes all necessary reagents and plates for analyzing S100B levels across multiple samples With minimal sample volume requirements and a streamlined workflow this assay kit enables efficient profiling of S100B which plays a role in diseases involving the central nervous system