Learn More
Meso Scale Discovery R-PLEX Human SCF ASYQP
SDP
Catalog No. 5000080197
Encompass
The R-Plex Human SCF Assay Kit from Meso Scale Discovery measures soluble stem cell factor (SCF) in human serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments with a typical assay time of 3 5 hours for 96 samples offering a sensitive high-throughput method for SCF measurement
Catalog No. 50-000-80197 Supplier Meso Scale Discovery Supplier No. K1510QR22
The R-Plex Human SCF Assay Kit from Meso Scale Discovery measures soluble stem cell factor (SCF) in human serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments with a typical assay time of 3 5 hours for 96 samples offering a sensitive high-throughput method for SCF measurement