Learn More
Meso Scale Discovery R-PLEX Human SCFR/Kit ASY
SDP
Catalog No. 5000078410
Encompass
The R-PLEX Human SCFR Kit Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human stem cell factor receptor (SCFR) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes all necessary reagents and calibrators offering high sensitivity a wide dynamic range and excellent performance in complex biological matrices enabling accurate quantification of human SCFR levels for life science research
Catalog No. 50-000-78410 Supplier Meso Scale Discovery Supplier No. K1515DR2
The R-PLEX Human SCFR Kit Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human stem cell factor receptor (SCFR) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes all necessary reagents and calibrators offering high sensitivity a wide dynamic range and excellent performance in complex biological matrices enabling accurate quantification of human SCFR levels for life science research