Learn More
Meso Scale Discovery R-PLEX Human SCFR/Kit ASYQP
SDP
Catalog No. 5000080558
Encompass
The R-PLEX Human SCFR Kit Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of human stem cell factor receptor (SCFR) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2500 pg/mL The total assay time is approximately 3 5 hours This product can aid in the study of SCFR levels related to diseases and treatments
Catalog No. 50-000-80558 Supplier Meso Scale Discovery Supplier No. K1515DR22
The R-PLEX Human SCFR Kit Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of human stem cell factor receptor (SCFR) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2500 pg/mL The total assay time is approximately 3 5 hours This product can aid in the study of SCFR levels related to diseases and treatments