Learn More
Meso Scale Discovery R-PLEX Human Sclerostin ASY
SDP
Catalog No. 5000080369
Encompass
The Meso Scale Discovery Sclerostin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of sclerostin in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This assay is intended for research use only and has not been tested for diagnostic use The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay
Catalog No. 50-000-80369 Supplier Meso Scale Discovery Supplier No. K151U4R2
The Meso Scale Discovery Sclerostin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of sclerostin in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements This assay is intended for research use only and has not been tested for diagnostic use The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay