Learn More
Meso Scale Discovery R-PLEX Human Sclerostin ASYQP
SDP
Catalog No. 5000081990
Encompass
The Meso Scale Discovery R-PLEX Human Sclerostin Assay is a multiplex immunoassay panel that measures the concentration of human Sclerostin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of Sclerostin which plays a key role in bone formation It can be used in biomarker research related to bone disorders and diseases The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81990 Supplier Meso Scale Discovery Supplier No. K151U4R22
The Meso Scale Discovery R-PLEX Human Sclerostin Assay is a multiplex immunoassay panel that measures the concentration of human Sclerostin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of Sclerostin which plays a key role in bone formation It can be used in biomarker research related to bone disorders and diseases The assay takes approximately 3 5 hours to complete