Learn More
Meso Scale Discovery R-PLEX Human SerpinA4 ASY
SDP
Catalog No. 5000079450
Encompass
The R-PLEX Human Serpin A4 Assay Kit from Meso Scale Discovery is used to quantitatively measure Serpin A4 protein levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample per well and takes approximately 3 5 hours to complete The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and sample diluent This product can aid research on Serpin A4 and its role in disease states such as cancer diabetes and inflammatory conditions
Catalog No. 50-000-79450 Supplier Meso Scale Discovery Supplier No. K151ABGR2
The R-PLEX Human Serpin A4 Assay Kit from Meso Scale Discovery is used to quantitatively measure Serpin A4 protein levels in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample per well and takes approximately 3 5 hours to complete The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and sample diluent This product can aid research on Serpin A4 and its role in disease states such as cancer diabetes and inflammatory conditions