Learn More
Meso Scale Discovery R-PLEX Human SerpinA4 ASYQP
SDP
Catalog No. 5000079490
Encompass
The R-PLEX Human Serpin A4 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure Serpin A4 protein levels in human serum plasma and cell culture supernatants It utilizes an electrochemiluminescent immunoassay method with high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls Sample incubation time is 2 hours The assay has a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL
Catalog No. 50-000-79490 Supplier Meso Scale Discovery Supplier No. K151ABGR22
The R-PLEX Human Serpin A4 Assay Kit from Meso Scale Diagnostics is used to quantitatively measure Serpin A4 protein levels in human serum plasma and cell culture supernatants It utilizes an electrochemiluminescent immunoassay method with high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls Sample incubation time is 2 hours The assay has a lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL