Learn More
Meso Scale Discovery R-PLEX Human SOD2 ASY
SDP
Catalog No. 5000080495
Encompass
The R-PLEX Human SOD2 Assay Kit from Meso Scale Discovery is used to quantitatively measure superoxide dismutase 2 (SOD2) in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent sandwich immunoassay method with high sensitivity and dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator SOD2 is measured in a range of 0 08-500 ng/mL with an incubation time of 2 hours This assay provides a simple rapid and robust tool to monitor SOD2 levels
Catalog No. 50-000-80495 Supplier Meso Scale Discovery Supplier No. K151AKWR2
The R-PLEX Human SOD2 Assay Kit from Meso Scale Discovery is used to quantitatively measure superoxide dismutase 2 (SOD2) in human serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent sandwich immunoassay method with high sensitivity and dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator SOD2 is measured in a range of 0 08-500 ng/mL with an incubation time of 2 hours This assay provides a simple rapid and robust tool to monitor SOD2 levels