Learn More
Meso Scale Discovery R-PLEX Human SOD2 ASYQP
SDP
Catalog No. 5000078865
Encompass
The R-PLEX Human SOD2 Assay Kit from Meso Scale Discovery measures superoxide dismutase 2 (SOD2) in human serum plasma cell culture supernatants and lysates It uses an electrochemiluminescent detection method with high sensitivity wide dynamic range and low sample volume The assay takes about 4 hours and includes reagents for one 96-well plate SOD2 is an enzyme that converts superoxide radicals into hydrogen peroxide and oxygen aiding in understanding oxidative stress This kit offers a fast sensitive and robust way to measure human SOD2
Catalog No. 50-000-78865 Supplier Meso Scale Discovery Supplier No. K151AKWR22
The R-PLEX Human SOD2 Assay Kit from Meso Scale Discovery measures superoxide dismutase 2 (SOD2) in human serum plasma cell culture supernatants and lysates It uses an electrochemiluminescent detection method with high sensitivity wide dynamic range and low sample volume The assay takes about 4 hours and includes reagents for one 96-well plate SOD2 is an enzyme that converts superoxide radicals into hydrogen peroxide and oxygen aiding in understanding oxidative stress This kit offers a fast sensitive and robust way to measure human SOD2