Learn More
Meso Scale Discovery R-PLEX Human SPINK-1 ASY
SDP
Catalog No. 5000080382
Encompass
The Meso Scale Discovery R-PLEX Human SPINK1 Assay is a multiplex immunoassay that measures SPINK1 protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less of sample it runs on QuickPlex instruments measuring SPINK1 from 0 11 to 10 000 pg/mL with a CV under 10% The assay completes in about 2 5 hours ensuring accurate reproducible SPINK1 quantification
Catalog No. 50-000-80382 Supplier Meso Scale Discovery Supplier No. K151A1R2
The Meso Scale Discovery R-PLEX Human SPINK1 Assay is a multiplex immunoassay that measures SPINK1 protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision Requiring 50 L or less of sample it runs on QuickPlex instruments measuring SPINK1 from 0 11 to 10 000 pg/mL with a CV under 10% The assay completes in about 2 5 hours ensuring accurate reproducible SPINK1 quantification