Learn More
Meso Scale Discovery R-PLEX Human SPINK-1 ASYQP
SDP
Catalog No. 5000079077
Encompass
The Meso Scale Discovery R-PLEX Human SPINK1 Assay is a multiplex immunoassay panel that measures SPINK1 protein levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The R-PLEX assay requires 50 L or less sample volume and can be fully automated on the MESO QuickPlex SQ 120 instrument The SPINK1 assay is intended for research use only and not for use in diagnostic procedures This product requires the MSD GOLD SULFO-TAG label and MSD read buffer for detection on the QuickPlex SQ 120
Catalog No. 50-000-79077 Supplier Meso Scale Discovery Supplier No. K151A1R22
The Meso Scale Discovery R-PLEX Human SPINK1 Assay is a multiplex immunoassay panel that measures SPINK1 protein levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The R-PLEX assay requires 50 L or less sample volume and can be fully automated on the MESO QuickPlex SQ 120 instrument The SPINK1 assay is intended for research use only and not for use in diagnostic procedures This product requires the MSD GOLD SULFO-TAG label and MSD read buffer for detection on the QuickPlex SQ 120