Learn More
Meso Scale Discovery R-PLEX Human SPK 1 ASY
SDP
Catalog No. 5000080480
Encompass
The R-PLEX Human SPK-1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures the levels of SPK-1 in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on MSD s QuickPlex SQ 120 instrument and MSD provides all the reagents required The R-PLEX assay allows for the quantitative measurement of SPK-1 which can aid in understanding its role in various biological processes and disease states
Catalog No. 50-000-80480 Supplier Meso Scale Discovery Supplier No. K151Z0R2
The R-PLEX Human SPK-1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures the levels of SPK-1 in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on MSD s QuickPlex SQ 120 instrument and MSD provides all the reagents required The R-PLEX assay allows for the quantitative measurement of SPK-1 which can aid in understanding its role in various biological processes and disease states