Learn More
Meso Scale Discovery R-PLEX Human SPK 1 ASYQP
SDP
Catalog No. 5000081842
Encompass
The Meso Scale Discovery Phospho (Ser/Thr) Akt 1/2/3 Assay Kit is used to quantitatively measure levels of Akt 1 Akt 2 and Akt 3 phosphorylated at serine and threonine residues This electrochemiluminescent assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer and calibrator The assay takes 3 5 hours to complete has a detection range of 50-50 000 pg/mL and shows minimal cross-reactivity with other proteins This product can help researchers study Akt phosphorylation and activation
Catalog No. 50-000-81842 Supplier Meso Scale Discovery Supplier No. K151Z0R22
The Meso Scale Discovery Phospho (Ser/Thr) Akt 1/2/3 Assay Kit is used to quantitatively measure levels of Akt 1 Akt 2 and Akt 3 phosphorylated at serine and threonine residues This electrochemiluminescent assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer and calibrator The assay takes 3 5 hours to complete has a detection range of 50-50 000 pg/mL and shows minimal cross-reactivity with other proteins This product can help researchers study Akt phosphorylation and activation