Learn More
Meso Scale Discovery R-PLEX Human Syndecan-4 ASY
SDP
Catalog No. 5000081980
Encompass
The R-PLEX Human Syndecan-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Syndecan-4 in serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of Syndecan-4 which plays a role in cell adhesion migration proliferation and angiogenesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration curve reagents and sample diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81980 Supplier Meso Scale Discovery Supplier No. K151AMGR2
Add to Cart
Add to Cart
The R-PLEX Human Syndecan-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Syndecan-4 in serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This assay allows for the quantitative measurement of Syndecan-4 which plays a role in cell adhesion migration proliferation and angiogenesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration curve reagents and sample diluent The assay takes approximately 3 5 hours to complete