Learn More
Meso Scale Discovery R-PLEX Human Syndecan-4 ASYQP
SDP
Catalog No. 5000082006
Encompass
The R-PLEX Human Syndecan-4 Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of human Syndecan-4 in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent (ECL) multiplex assay uses MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of Syndecan-4 on the Meso Scale Discovery platform The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and assay diluent The assay has a typical sensitivity of 19 pg/mL and an upper limit of quantification of 50 000 pg/mL
Catalog No. 50-000-82006 Supplier Meso Scale Discovery Supplier No. K151AMGR22
The R-PLEX Human Syndecan-4 Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of human Syndecan-4 in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent (ECL) multiplex assay uses MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of Syndecan-4 on the Meso Scale Discovery platform The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and assay diluent The assay has a typical sensitivity of 19 pg/mL and an upper limit of quantification of 50 000 pg/mL