Learn More
Meso Scale Discovery R-PLEX Human Tau total ASY
SDP
Catalog No. 5000081571
Encompass
The Meso Scale Discovery R-PLEX Human Tau Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF It uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible assay capable of detecting tau levels as low as 50 pg/mL with an upper limit of quantification of 25 000 pg/mL The assay has high specificity for both normal and phosphorylated forms of tau Each kit provides sufficient reagents to assay one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-81571 Supplier Meso Scale Discovery Supplier No. K151AGTR2
The Meso Scale Discovery R-PLEX Human Tau Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF It uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible assay capable of detecting tau levels as low as 50 pg/mL with an upper limit of quantification of 25 000 pg/mL The assay has high specificity for both normal and phosphorylated forms of tau Each kit provides sufficient reagents to assay one 96-well plate testing 40 samples in duplicate