Learn More
Meso Scale Discovery R-PLEX Human Tau total ASYQP
SDP
Catalog No. 5000081572
Encompass
The Meso Scale Discovery R-Plex Human Tau Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF It uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of tau levels The assay has a detection range of 50-25 000 pg/mL and exhibits no cross-reactivity with other proteins It requires 50 L of sample and can be read on MSD instrument platforms The R-Plex assay enables high-throughput analysis of tau a biomarker associated with Alzheimer s disease and other neurodegenerative disorders
Catalog No. 50-000-81572 Supplier Meso Scale Discovery Supplier No. K151AGTR22
The Meso Scale Discovery R-Plex Human Tau Total Assay is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF It uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid quantification of tau levels The assay has a detection range of 50-25 000 pg/mL and exhibits no cross-reactivity with other proteins It requires 50 L of sample and can be read on MSD instrument platforms The R-Plex assay enables high-throughput analysis of tau a biomarker associated with Alzheimer s disease and other neurodegenerative disorders