Learn More
Meso Scale Discovery R-PLEX Human Tenascin C ASY
SDP
Catalog No. 5000079726
Encompass
The R-PLEX Human Tenascin-C Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of Tenascin-C in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of Tenascin-C an extracellular matrix glycoprotein implicated in tissue injury and cancer progression With a detection range of 1 56-400 ng/mL and minimal required sample volume this assay allows high-throughput and accurate quantification of Tenascin-C levels from small sample sizes
Catalog No. 50-000-79726 Supplier Meso Scale Discovery Supplier No. K151ZQR2
The R-PLEX Human Tenascin-C Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of Tenascin-C in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of Tenascin-C an extracellular matrix glycoprotein implicated in tissue injury and cancer progression With a detection range of 1 56-400 ng/mL and minimal required sample volume this assay allows high-throughput and accurate quantification of Tenascin-C levels from small sample sizes