Learn More
Meso Scale Discovery R-PLEX Human Tenascin C ASYQP
SDP
Catalog No. 5000080486
Encompass
The R-PLEX Human Tenascin-C Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of Tenascin-C in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The assay has a detection range of 0 05-1000 ng/mL and requires only 50 L sample volume per well The R-PLEX assay format allows for the simultaneous measurement of multiple analytes from a single sample This assay can be used to aid in the diagnosis and monitoring of diseases associated with changes in Tenascin-C expression
Catalog No. 50-000-80486 Supplier Meso Scale Discovery Supplier No. K151ZQR22
The R-PLEX Human Tenascin-C Assay from Meso Scale Discovery is an electrochemiluminescent immunoassay designed for the quantitative measurement of Tenascin-C in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The assay has a detection range of 0 05-1000 ng/mL and requires only 50 L sample volume per well The R-PLEX assay format allows for the simultaneous measurement of multiple analytes from a single sample This assay can be used to aid in the diagnosis and monitoring of diseases associated with changes in Tenascin-C expression