Learn More
Meso Scale Discovery R-PLEX Human TFF2 ASY
SDP
Catalog No. 5000078945
Encompass
The R-PLEX Human TFF2 Assay from Meso Scale Diagnostics is a multiplex immunoassay measuring Trefoil Factor 2 (TFF2) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity up to 8-logs dynamic range and low sample volume The all-in-one kit includes plates with capture antibody detection antibody read buffer calibrator and controls It quantifies TFF2 important for mucosal protection and healing aiding the study of gastrointestinal diseases
Catalog No. 50-000-78945 Supplier Meso Scale Discovery Supplier No. K151ACMR2
The R-PLEX Human TFF2 Assay from Meso Scale Diagnostics is a multiplex immunoassay measuring Trefoil Factor 2 (TFF2) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity up to 8-logs dynamic range and low sample volume The all-in-one kit includes plates with capture antibody detection antibody read buffer calibrator and controls It quantifies TFF2 important for mucosal protection and healing aiding the study of gastrointestinal diseases