Learn More
Meso Scale Discovery R-PLEX Human TFF2 ASYQP
SDP
Catalog No. 5000079579
Encompass
The R-PLEX Human TFF2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Trefoil Factor 2 (TFF2) in human serum plasma saliva milk and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators and reagents required to perform testing of 46 samples in duplicate It can aid in the research of gastric mucosa protection and repair mechanisms
Catalog No. 50-000-79579 Supplier Meso Scale Discovery Supplier No. K151ACMR22
The R-PLEX Human TFF2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Trefoil Factor 2 (TFF2) in human serum plasma saliva milk and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators and reagents required to perform testing of 46 samples in duplicate It can aid in the research of gastric mucosa protection and repair mechanisms