Learn More
Meso Scale Discovery R-PLEX Human TGF- ASY
SDP
Catalog No. 5000079534
Encompass
The R-Plex Human TGF-alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures the cytokine TGF-alpha in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range The assay requires 50 L of sample and can be run in as little as 3 5 hours The lower limit of detection is 1 15 pg/mL This assay allows for the quantitative measurement of TGF-alpha across a range of biological research applications
Catalog No. 50-000-79534 Supplier Meso Scale Discovery Supplier No. K151ACNR2
The R-Plex Human TGF-alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures the cytokine TGF-alpha in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range The assay requires 50 L of sample and can be run in as little as 3 5 hours The lower limit of detection is 1 15 pg/mL This assay allows for the quantitative measurement of TGF-alpha across a range of biological research applications