Learn More
Meso Scale Discovery R-PLEX Human TGF- ASYQP
SDP
Catalog No. 5000079505
Encompass
The R-PLEX Human TGF-alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures TGF-alpha levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a broad dynamic range The assay has high specificity and low cross-reactivity Each kit provides reagents for one 96-well plate which can assay 46 samples in duplicate The lower limit of detection is 1 15 pg/mL This quantitative immunoassay provides accurate and reproducible results for measuring TGF-alpha in biological samples
Catalog No. 50-000-79505 Supplier Meso Scale Discovery Supplier No. K151ACNR22
The R-PLEX Human TGF-alpha Assay from Meso Scale Discovery is a multiplex immunoassay that measures TGF-alpha levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a broad dynamic range The assay has high specificity and low cross-reactivity Each kit provides reagents for one 96-well plate which can assay 46 samples in duplicate The lower limit of detection is 1 15 pg/mL This quantitative immunoassay provides accurate and reproducible results for measuring TGF-alpha in biological samples