Learn More
Meso Scale Discovery R-PLEX Human TGF-B RII ASY
SDP
Catalog No. 5000078920
Encompass
The R-PLEX Human TGF-beta RII Assay is a multiplex immunoassay kit for measuring TGF-beta receptor type II in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture and detection antibodies standards controls buffers substrate and consumables It quantifies TGF-beta RII with high precision and accuracy compatible with common samples providing results in under 3 hours It aids in studying TGF-beta signaling in cancer fibrosis immunology and stem cell research
Catalog No. 50-000-78920 Supplier Meso Scale Discovery Supplier No. K151ACYR2
The R-PLEX Human TGF-beta RII Assay is a multiplex immunoassay kit for measuring TGF-beta receptor type II in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture and detection antibodies standards controls buffers substrate and consumables It quantifies TGF-beta RII with high precision and accuracy compatible with common samples providing results in under 3 hours It aids in studying TGF-beta signaling in cancer fibrosis immunology and stem cell research