Learn More
Meso Scale Discovery R-PLEX Human TGF-B RII ASYQP
SDP
Catalog No. 5000079031
Encompass
The R-Plex Human TGF-beta RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TGF-beta RII in cell and tissue lysates It utilizes electrochemiluminescence (ECL) detection technology and patterned arrays This assay kit contains all necessary reagents and materials to run the immunoassay on SECTOR Imager instrument platforms It allows high-throughput sensitive and reproducible quantification of TGF-beta RII from small sample volumes with minimal sample dilution The kit includes assay buffer detection antibody wash buffer readout buffer calibrator blend serum diluent and plate
Catalog No. 50-000-79031 Supplier Meso Scale Discovery Supplier No. K151ACYR22
The R-Plex Human TGF-beta RII Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TGF-beta RII in cell and tissue lysates It utilizes electrochemiluminescence (ECL) detection technology and patterned arrays This assay kit contains all necessary reagents and materials to run the immunoassay on SECTOR Imager instrument platforms It allows high-throughput sensitive and reproducible quantification of TGF-beta RII from small sample volumes with minimal sample dilution The kit includes assay buffer detection antibody wash buffer readout buffer calibrator blend serum diluent and plate