Learn More
Meso Scale Discovery R-PLEX Human TGM-2 ASYQP
SDP
Catalog No. 5000078866
Encompass
The R-PLEX Human TGM-2 Assay Kit from Meso Scale Discovery measures tissue transglutaminase (TGM-2) in human serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours aiding in understanding TGM-2 levels in various diseases
Catalog No. 50-000-78866 Supplier Meso Scale Discovery Supplier No. K151AKCR22
The R-PLEX Human TGM-2 Assay Kit from Meso Scale Discovery measures tissue transglutaminase (TGM-2) in human serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours aiding in understanding TGM-2 levels in various diseases