Learn More
Meso Scale Discovery R-PLEX Human TIMP-1 ASYQP
SDP
Catalog No. 5000081802
Encompass
The Meso Scale Discovery R-PLEX Human TIMP-1 Assay is a multiplex immunoassay panel that measures TIMP-1 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology on patterned arrays for high sensitivity and a wide dynamic range The assay requires 50 L or less of sample and can be read on the MESO QuickPlex SQ 120 instrument The assay is available as a singleplex or as part of a multiplex panel This assay allows for the quantitative measurement of TIMP-1 which can aid in the study of tissue remodeling inflammation and disease progression
Catalog No. 50-000-81802 Supplier Meso Scale Discovery Supplier No. K151YOR22
The Meso Scale Discovery R-PLEX Human TIMP-1 Assay is a multiplex immunoassay panel that measures TIMP-1 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology on patterned arrays for high sensitivity and a wide dynamic range The assay requires 50 L or less of sample and can be read on the MESO QuickPlex SQ 120 instrument The assay is available as a singleplex or as part of a multiplex panel This assay allows for the quantitative measurement of TIMP-1 which can aid in the study of tissue remodeling inflammation and disease progression