Learn More
Meso Scale Discovery R-PLEX Human TIMP-4 ASY
SDP
Catalog No. 5000079650
Encompass
The Meso Scale Discovery R-PLEX Human TIMP-4 Assay is a multiplex immunoassay panel that measures TIMP-4 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify TIMP-4 concentrations between 0 03 and 10 000 pg/mL It utilizes a sandwich immunoassay format with SULFO-TAG labels that emit light upon electrochemical stimulation This 6-spot 96-well configuration allows rapid and efficient analysis of up to 80 samples in duplicate
Catalog No. 50-000-79650 Supplier Meso Scale Discovery Supplier No. K151AMJR2
The Meso Scale Discovery R-PLEX Human TIMP-4 Assay is a multiplex immunoassay panel that measures TIMP-4 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can quantify TIMP-4 concentrations between 0 03 and 10 000 pg/mL It utilizes a sandwich immunoassay format with SULFO-TAG labels that emit light upon electrochemical stimulation This 6-spot 96-well configuration allows rapid and efficient analysis of up to 80 samples in duplicate