Learn More
Meso Scale Discovery R-PLEX Human TIMP-4 ASYQP
SDP
Catalog No. 5000082437
Encompass
The Meso Scale Discovery R-PLEX Human TIMP-4 Assay is a multiplex immunoassay that measures TIMP-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay quantifies TIMP-4 from 0 03 to 10 000 pg/mL using a sandwich immunoassay format with SULFO-TAG labels This 6-spot 96-well assay allows simultaneous measurement of TIMP-4 and other biomarkers providing reliable data from small samples in a few hours
Catalog No. 50-000-82437 Supplier Meso Scale Discovery Supplier No. K151AMJR22
The Meso Scale Discovery R-PLEX Human TIMP-4 Assay is a multiplex immunoassay that measures TIMP-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay quantifies TIMP-4 from 0 03 to 10 000 pg/mL using a sandwich immunoassay format with SULFO-TAG labels This 6-spot 96-well assay allows simultaneous measurement of TIMP-4 and other biomarkers providing reliable data from small samples in a few hours