Learn More
Meso Scale Discovery R-PLEX Human Trk-A total ASY
SDP
Catalog No. 5000079608
Encompass
The R-PLEX Human TrkA Total Assay Kit from Meso Scale Discovery measures total TrkA in human cell and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal matrix interference The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay accurately quantifies total TrkA levels
Catalog No. 50-000-79608 Supplier Meso Scale Discovery Supplier No. K151AKXR2
The R-PLEX Human TrkA Total Assay Kit from Meso Scale Discovery measures total TrkA in human cell and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and minimal matrix interference The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay accurately quantifies total TrkA levels