Learn More
Meso Scale Discovery R-PLEX Human TSP-1 ASYQP
SDP
Catalog No. 5000079530
Encompass
The R-PLEX Human TSP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of Thrombospondin-1 (TSP-1) in human serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator This quantitative immunoassay allows the user to accurately determine TSP-1 levels across multiple samples
Catalog No. 50-000-79530 Supplier Meso Scale Discovery Supplier No. K1519FR22
The R-PLEX Human TSP-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of Thrombospondin-1 (TSP-1) in human serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator This quantitative immunoassay allows the user to accurately determine TSP-1 levels across multiple samples