Learn More
Meso Scale Discovery R-PLEX Human TSP-2 ASY
SDP
Catalog No. 5000082468
Encompass
The R-PLEX Human TSP-2 Assay Kit from Meso Scale Discovery measures Thrombospondin-2 in human serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls buffers and seals It quantifies TSP-2 concentrations from 0 3-10 000 pg/mL with precision aiding in the study of tissue remodeling and angiogenesis
Catalog No. 50-000-82468 Supplier Meso Scale Discovery Supplier No. K151ALGR2
The R-PLEX Human TSP-2 Assay Kit from Meso Scale Discovery measures Thrombospondin-2 in human serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls buffers and seals It quantifies TSP-2 concentrations from 0 3-10 000 pg/mL with precision aiding in the study of tissue remodeling and angiogenesis