Learn More
Meso Scale Discovery R-PLEX Human TWEAK/TNFSF12 ASY
SDP
Catalog No. 5000082271
Encompass
The R-PLEX Human TWEAK (TNFSF12) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TWEAK (TNF-related weak inducer of apoptosis) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate pre-coated with capture antibody a detection antibody and calibrator It can detect TWEAK concentrations as low as 17 pg/mL with an approximate 3 5 hour assay time This assay is for research use only
Catalog No. 50-000-82271 Supplier Meso Scale Discovery Supplier No. K151P5R2
Add to Cart
Add to Cart
The R-PLEX Human TWEAK (TNFSF12) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures TWEAK (TNF-related weak inducer of apoptosis) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate pre-coated with capture antibody a detection antibody and calibrator It can detect TWEAK concentrations as low as 17 pg/mL with an approximate 3 5 hour assay time This assay is for research use only