Learn More
Meso Scale Discovery R-PLEX Human TYRO-3 ASY
SDP
Catalog No. 5000082062
Encompass
The R-PLEX Human Tyro-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human Tyro-3 protein in serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay has a typical lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL for human Tyro-3
Catalog No. 50-000-82062 Supplier Meso Scale Discovery Supplier No. K151AMKR2
The R-PLEX Human Tyro-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human Tyro-3 protein in serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay has a typical lower limit of detection of 19 pg/mL and an upper limit of 100 000 pg/mL for human Tyro-3