Learn More
Meso Scale Discovery R-PLEX Human TYRO-3 ASYQP
SDP
Catalog No. 5000082438
Encompass
The R-PLEX Human Tyro-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human Tyro-3 protein in serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay can detect Tyro-3 concentrations as low as 17 pg/mL with an upper limit of 100 000 pg/mL
Catalog No. 50-000-82438 Supplier Meso Scale Discovery Supplier No. K151AMKR22
The R-PLEX Human Tyro-3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human Tyro-3 protein in serum plasma and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent The assay can detect Tyro-3 concentrations as low as 17 pg/mL with an upper limit of 100 000 pg/mL