Learn More
Meso Scale Discovery R-PLEX Human U-PAR ASY
SDP
Catalog No. 5000080604
Encompass
The R-PLEX Human U-PLEX Assay is a multiplex immunoassay panel that measures 38 biomarkers related to inflammation immune response and tissue remodeling It utilizes electrochemiluminescence detection technology on the Meso Scale Discovery platform The assay requires 50 L of sample and can be used with serum plasma and cell culture supernatants It has high sensitivity broad dynamic range and requires low sample volume The panel includes cytokines chemokines growth factors soluble receptors and matrix metalloproteinases It is designed for applications in immunology oncology neurobiology cardiovascular and metabolic research
Catalog No. 50-000-80604 Supplier Meso Scale Discovery Supplier No. K1514SR2
The R-PLEX Human U-PLEX Assay is a multiplex immunoassay panel that measures 38 biomarkers related to inflammation immune response and tissue remodeling It utilizes electrochemiluminescence detection technology on the Meso Scale Discovery platform The assay requires 50 L of sample and can be used with serum plasma and cell culture supernatants It has high sensitivity broad dynamic range and requires low sample volume The panel includes cytokines chemokines growth factors soluble receptors and matrix metalloproteinases It is designed for applications in immunology oncology neurobiology cardiovascular and metabolic research