Learn More
Meso Scale Discovery R-PLEX Human U-PAR ASYQP
SDP
Catalog No. 5000080605
Encompass
The R-PLEX Human U-PLEX Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures the concentrations of 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN- TNF- IL-8 IP-10 MCP-1) in serum plasma and cell culture supernatants This electrochemiluminescence assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels to provide sensitive and reproducible results from a small sample volume The 10-plex panel allows efficient quantification of multiple analytes from a single sample
Catalog No. 50-000-80605 Supplier Meso Scale Discovery Supplier No. K1514SR22
The R-PLEX Human U-PLEX Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures the concentrations of 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN- TNF- IL-8 IP-10 MCP-1) in serum plasma and cell culture supernatants This electrochemiluminescence assay utilizes patterned arrays and MSD s proprietary SULFO-TAG labels to provide sensitive and reproducible results from a small sample volume The 10-plex panel allows efficient quantification of multiple analytes from a single sample