Learn More
Meso Scale Discovery R-PLEX Human VE-Cadherin ASY
SDP
Catalog No. 5000078219
Encompass
The R-Plex Human VE-Cadherin Assay is a multiplex immunoassay kit used to measure vascular endothelial (VE)-cadherin in human serum plasma and cell culture supernatants It utilizes a quantitative sandwich immunoassay technique and can measure VE-cadherin concentrations between 0 313 ng/mL to 20 ng/mL The assay requires a low sample volume of 50 L and can be read on commonly available Luminex-based readers It includes a 96-well plate pre-coated with analyte capture antibodies detection antibodies standards controls assay and sample diluents wash buffer streptavidin-phycoerythrin and a plate seal The assay takes around 3 hours to complete
Catalog No. 50-000-78219 Supplier Meso Scale Discovery Supplier No. K151AJVR2
The R-Plex Human VE-Cadherin Assay is a multiplex immunoassay kit used to measure vascular endothelial (VE)-cadherin in human serum plasma and cell culture supernatants It utilizes a quantitative sandwich immunoassay technique and can measure VE-cadherin concentrations between 0 313 ng/mL to 20 ng/mL The assay requires a low sample volume of 50 L and can be read on commonly available Luminex-based readers It includes a 96-well plate pre-coated with analyte capture antibodies detection antibodies standards controls assay and sample diluents wash buffer streptavidin-phycoerythrin and a plate seal The assay takes around 3 hours to complete